Prostaglandin F2α tris salt 98%

Supplier: Ambeed
Danger

Synonyms: Dinoprost tromethamine salt, 2-Amino-2-(hydroxymethyl)propane-1,3-diol (Z)-7-((1R,2R,3R,5S)-3,5-Dihydroxy-2-((S,E)-3-hydroxyoct-1-en-1-yl)cyclopentyl)hept-5-enoate

A385706-10MG A385706-1MG A385706-5MG
76724-926EA 21.22 USD
76724-926 76724-928 76724-930
Prostaglandin F2α tris salt 98%
Prostaglandin F2alpha tris salt

Precaución: for Research Use Only

Formula: C₂₄H₄₅NO₈
MW: 475.623 g/mol
MDL Number: MFCD00077863
CAS Number: 38562-01-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR