Phenylglyoxal monohydrate 95%

Supplier: Ambeed
Warning

Synonyms: 2,2-Dihydroxy-1-phenylethan-1-one, Benzoylformaldehyde, Phenylglyoxal hydrate, 2,2-Dihydroxyacetophenone

A385200-10G A385200-1G A385200-25MG A385200-5G A385200-250MG A385200-25G A385200-100G
76724-908EA 49 USD
76724-908 76724-910 76724-912 76724-914 77594-166 77668-792 77879-829
Phenylglyoxal monohydrate 95%
Phenylglyoxal monohydrate

Precaución: for Research Use Only

Formula: C₈H₆O₂·H₂O
MDL Number: MFCD00149499
CAS Number: 1075-06-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR