Potassium tetrachloroaurate 98%

Supplier: Ambeed
Warning

Synonyms: Gold potassium chloride, Potassium tetrachloroaurate(III), Potassium gold(III) chloride

A382520-100MG A382520-1G A382520-250MG A382520-5G
76724-886EA 13.8 USD
76724-886 76724-888 76724-890 76724-892
Potassium tetrachloroaurate 98%
Potassium tetrachloroaurate

Precaución: for Research Use Only

Formula: AuCl₄K
MW: 377.88 g/mol
MDL Number: MFCD00043088
CAS Number: 13682-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR