Potassium-3-carboxyphenyltrifluoroborate 98%

Supplier: Ambeed
Warning

A381211-1G A381211-250MG A381211-5G
76724-880EA 64 USD
76724-880 76724-882 76724-884
Potassium-3-carboxyphenyltrifluoroborate 98%
Potassium 3-carboxyphenyltrifluoroborate

Precaución: for Research Use Only

Formula: C₇H₅BF₃KO₂
MDL Number: MFCD07784423
CAS Number: 850313-91-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR