Potassium (2,6-dimethylphenyl)trifluoroborate 97%

Supplier: Ambeed
Warning

Synonyms: Potassium 2,6-dimethylphenyltrifluoroborate

A380954-1G A380954-250MG A380954-5G
76724-874EA 37 USD
76724-874 76724-876 76724-878
Potassium (2,6-dimethylphenyl)trifluoroborate 97%
Potassium (2,6-dimethylphenyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₈H₉BF₃K
MW: 212.06 g/mol
MDL Number: MFCD04974103
CAS Number: 561328-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR