Sorbic acid ≥99%

Supplier: Ambeed
Warning

Synonyms: (E,E)-2,4-Hexadienoic acid, (2E,4E)-2,4-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, 2,4-Hexadienoic acid, (2E,4E)-Hexa-2,4-dienoic acid

A379771-1000G A379771-100G A379771-25G A379771-500G
76724-856EA 33.95 USD
76724-856 76724-858 76724-860 76724-862
Sorbic acid ≥99%
Sorbic acid

Precaución: for Research Use Only

Formula: CH₃CH=CHCH=CHCOOH
MW: 112.13 g/mol
MDL Number: MFCD00002703
CAS Number: 110-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR