[3-(6-Aminohexylamino)propyl]trimethoxysilane 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
N1-[3-(Trimethoxysilyl)propyl]hexane-1,6-diamine, N-[3-(Trimethoxysilyl)propyl]hexamethylenediamine
A377819-1G
A377819-250MG
A377819-5G
76724-850EA
29.71
USD
76724-850
76724-852
76724-854
[3-(6-Aminohexylamino)propyl]trimethoxysilane 97%
[3-(6-Aminohexylamino)propyl]trimethoxysilane
Precaución: for Research Use Only
Formula:
C₁₂H₃₀N₂O₃Si MW: 278.47 g/mol |
MDL Number:
MFCD00054943 CAS Number: 51895-58-0 UN: 1760 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...