(3S,4R,5S)-Methyl 3,4,5,6-tetrahydroxy-2-oxohexanoate 95%

Supplier: Ambeed
Warning

A377418-100MG A377418-1G A377418-250MG A377418-5G
76724-842EA 54 USD
76724-842 76724-844 76724-846 76724-848
(3S,4R,5S)-Methyl 3,4,5,6-tetrahydroxy-2-oxohexanoate 95%
(3S,4R,5S)-Methyl 3,4,5,6-tetrahydroxy-2-oxohexanoate

Precaución: for Research Use Only

Formula: C₇H₁₂O₇
MDL Number: MFCD00021987
CAS Number: 3031-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR