(S)-2-((tert-Butoxycarbonyl)amino)-2-(4-chlorophenyl)acetic acid 97%

Supplier: Ambeed
Warning

A376560-100MG A376560-1G A376560-250MG
76724-832EA 49.87 USD
76724-832 76724-834 76724-836
(S)-2-((tert-Butoxycarbonyl)amino)-2-(4-chlorophenyl)acetic acid 97%
(S)-2-((tert-Butoxycarbonyl)amino)-2-(4-chlorophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₆ClNO₄
MW: 285.72 g/mol
MDL Number: MFCD07371721
CAS Number: 917027-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR