Phenyl acridine-9-carboxylate 95%

Supplier: Ambeed
Warning

A376366-100MG A376366-1G A376366-250MG
76724-826EA 27 USD
76724-826 76724-828 76724-830
Phenyl acridine-9-carboxylate 95%
Phenyl acridine-9-carboxylate

Precaución: for Research Use Only

Formula: C₂₀H₁₃NO₂
MDL Number: MFCD00075288
CAS Number: 109392-90-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR