Ethyl N-[4-(trifluoromethyl)pyrimid-2-yl]piperidine-4-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 1-(4-(trifluoromethyl)pyrimidin-2-yl)piperidine-4-carboxylate

A375280-100MG A375280-1G
76724-818EA 31.83 USD
76724-818 76724-820
Ethyl N-[4-(trifluoromethyl)pyrimid-2-yl]piperidine-4-carboxylate 98%
Ethyl N-[4-(trifluoromethyl)pyrimid-2-yl]piperidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₆F₃N₃O₂
MW: 303.28 g/mol
MDL Number: MFCD00661832
CAS Number: 215654-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR