Octaethylene glycol 97%

Supplier: Ambeed
Warning

Synonyms: PEG-08, 3,6,9,12,15,18,21-Heptaoxatricosane-1,23-diol

A374820-100G A374820-100MG A374820-10G A374820-1G A374820-250MG A374820-25G A374820-5G
76724-804EA 1094.85 USD
76724-804 76724-806 76724-808 76724-810 76724-812 76724-814 76724-816
Octaethylene glycol 97%
Octaethylene glycol

Precaución: for Research Use Only

Formula: C₁₆H₃₄O₉
MW: 370.44 g/mol
MDL Number: MFCD00698694
CAS Number: 5117-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR