N-[2-(Dimethylamino)ethyl]-N-methylformamide 98%

Supplier: Ambeed
Warning

Synonyms: N-(2-(Dimethylamino)ethyl)-N-methylformamide, N-Formyl-N,N',N'-trimethylethylenediamine

A374653-1G A374653-5G
76724-800EA 9.55 USD
76724-800 76724-802
N-[2-(Dimethylamino)ethyl]-N-methylformamide 98%
N-[2-(Dimethylamino)ethyl]-N-methylformamide

Precaución: for Research Use Only

Formula: C₆H₁₄N₂O
MW: 130.19 g/mol
MDL Number: MFCD04039895
CAS Number: 105669-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR