Potassium-3-(methylthio)phenyltrifluoroborate 98%

Supplier: Ambeed
Warning

A374305-1G A374305-25G A374305-5G
76724-794EA 12.74 USD
76724-794 76724-796 76724-798
Potassium-3-(methylthio)phenyltrifluoroborate 98%
Potassium 3-(methylthio)phenyltrifluoroborate

Precaución: for Research Use Only

Formula: C₇H₇BF₃KS
MW: 230.1 g/mol
MDL Number: MFCD04115747
CAS Number: 850623-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR