1,3,3-Trimethyl-2-methyleneindoline 98%

Supplier: Ambeed
Warning

A374094-25G A374094-500G A374094-100G A374094-5G
76724-790EA 22 USD
76724-790 76724-792 77572-118 77879-817
1,3,3-Trimethyl-2-methyleneindoline 98%
1,3,3-Trimethyl-2-methyleneindoline

Precaución: for Research Use Only

Formula: C₁₂H₁₅N
MDL Number: MFCD00005813
CAS Number: 118-12-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR