Methyl 4-bromo-3,5-dimethoxybenzoate 97%

Supplier: Ambeed
Warning

A371426-100G A371426-1G A371426-25G A371426-5G
76724-768EA 98.67 USD
76724-768 76724-770 76724-772 76724-774
Methyl 4-bromo-3,5-dimethoxybenzoate 97%
Methyl 4-bromo-3,5-dimethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₄
MW: 275.096 g/mol
MDL Number: MFCD06628784
CAS Number: 26050-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR