Nimesulide 98%

Supplier: Ambeed
Danger

Synonyms: N-(4-Nitro-2-phenoxyphenyl)methanesulphonamide

A370794-100MG A370794-10G A370794-1G A370794-25MG A370794-5G A370794-1MG A370794-25G
76724-758EA 8.49 USD
76724-758 76724-760 76724-762 76724-764 76724-766 77601-640 77668-716
Nimesulide 98%
Nimesulide

Precaución: for Research Use Only

Formula: C₁₃H₁₂N₂O₅S
MW: 308.308 g/mol
MDL Number: MFCD00079470
CAS Number: 51803-78-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR