Sodium 2-(hydroxymethylamino)acetate 90%

Supplier: Ambeed
Warning

A370320-100MG A370320-250MG A370320-5G
76724-752EA 36 USD
76724-752 76724-754 76724-756
Sodium 2-(hydroxymethylamino)acetate 90%
Sodium 2-(hydroxymethylamino)acetate

Precaución: for Research Use Only

Formula: C₃H₆NO₃·Na
MDL Number: MFCD04307769
CAS Number: 70161-44-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR