4-Nitrophenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 4,4,5,5-Tetramethyl-2-(4-nitrophenyl)-1,3,2-dioxaborolane, 4-Nitrophenylboronic acid Pinacol ester, 1-Nitro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 4-Nitrobenzeneboronic acid pinacol ester

A368897-100G A368897-10G A368897-1G A368897-250MG A368897-25G A368897-5G
76724-726EA 256 USD
76724-726 76724-728 76724-730 76724-732 76724-734 76724-736
4-Nitrophenylboronic acid pinacol ester 98%
4-Nitrophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₆BNO₄
MDL Number: MFCD02179437
CAS Number: 171364-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR