Bis(2,4-pentanedionato)molybdenum(VI) dioxide 98% stabilised

Supplier: Ambeed
Warning

Synonyms: Molybdenum(VI)dioxy Acetylacetonate, Acetylacetone Molybdenum(VI)dioxy Salt, Molybdenum(VI) oxide bis(acetylacetonate), Molybdenum(VI) dioxide bis(acetylacetonate), Molybdenyl acetylacetonate, Bis(acetylacetonato)dioxomolybdenum(VI)

Stabilization: Stabilized with acetylacetone 1~2%

A368833-5G A368833-25G A368833-100G A368833-500G
76724-724EA 6 USD
76724-724 77601-628 77879-796 77879-797
Bis(2,4-pentanedionato)molybdenum(VI) dioxide 98% stabilised
Bis(2,4-pentanedionato)molybdenum(VI) dioxide

Precaución: for Research Use Only

Formula: C₁₀H₁₄MoO₆
MDL Number: MFCD00011506
CAS Number: 17524-05-9
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR