Nicotinanilide 98%

Supplier: Ambeed
Warning

A368109-1G A368109-25G A368109-5G
76724-710EA 23 USD
76724-710 76724-712 76724-714
Nicotinanilide 98%
Nicotinanilide

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₂O
MDL Number: MFCD00023572
CAS Number: 1752-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR