Ocaperidone 97%

Supplier: Ambeed
Warning

Synonyms: 3-[2-[4-(6-Fluorobenzo[d]isoxazol-3-yl)piperidin-1-yl]ethyl]-2,9-dimethyl-4H-pyrido[1,2-a]pyrimidin-4-one

A367930-10MG A367930-1MG A367930-25MG A367930-50MG A367930-5MG
76724-700EA 115.64 USD
76724-700 76724-702 76724-704 76724-706 76724-708
Ocaperidone 97%
Ocaperidone

Precaución: for Research Use Only

Formula: C₂₄H₂₅FN₄O₂
MW: 420.49 g/mol
MDL Number: MFCD00866668
CAS Number: 129029-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR