Potassium hydrogen carbonate 99%

Supplier: Ambeed
Warning

A367522-1000G A367522-100G A367522-500G
76724-694EA 11.67 USD
76724-694 76724-696 76724-698
Potassium hydrogen carbonate 99%
Potassium hydrogen carbonate

Precaución: for Research Use Only

Formula: KHCO₃
MDL Number: MFCD00011402
CAS Number: 298-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR