Sarcosine sodium salt ∼ 35% in water

Supplier: Ambeed
Warning

Synonyms: Sodium Sarcosinate, N-Methylglycine Sodium Salt, Me-Gly-ONa, (Methylamino)acetic Acid Sodium Salt, Sodium (Methylamino)acetate

A366212-5G A366212-100G A366212-500G
76724-692EA 8 USD
76724-692 77872-561 77872-562
Sarcosine sodium salt ∼ 35% in water
Sarcosine sodium salt

Precaución: for Research Use Only

Formula: C₃H₇NO₂
MW: 111.08 g/mol
MDL Number: MFCD00068282
CAS Number: 4316-73-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR