1,3-Di-tert-butylthiourea 98%

Supplier: Ambeed
Warning

A366042-25G A366042-5G
76724-688EA 60.48 USD
76724-688 76724-690
1,3-Di-tert-butylthiourea 98%
1,3-Di-tert-butylthiourea

Precaución: for Research Use Only

Formula: C₉H₂₀N₂S
MW: 188.33 g/mol
MDL Number: MFCD00026272
CAS Number: 4041-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR