Propargyl acetate 95%

Supplier: Ambeed
Danger

Synonyms: Propargylacetate

A365876-1G A365876-5G A365876-25G
76724-684EA 11 USD
76724-684 76724-686 77879-789
Propargyl acetate 95%
Propargyl acetate

Precaución: for Research Use Only

Formula: C₅H₆O₂
MW: 98.10 g/mol
MDL Number: MFCD00041601
CAS Number: 627-09-8
UN: 1992
ADR: 3,6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR