4-(3-Hydroxyprop-1-en-1-yl)-2,6-dimethoxyphenol 97% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A365753-100MG A365753-1G A365753-250MG A365753-5G
76724-682EA 97 USD
76724-682 77572-086 77668-694 77872-560
4-(3-Hydroxyprop-1-en-1-yl)-2,6-dimethoxyphenol 97% stabilized
4-(3-Hydroxyprop-1-en-1-yl)-2,6-dimethoxyphenol

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₄
MDL Number: MFCD00192441
CAS Number: 537-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR