Potassium(2-methylphenyl)trifluoroborate 96%

Supplier: Ambeed
Warning

Synonyms: (2-Methylphenyl)trifluoroboric acid potassium salt, Potassium trifluoro(o-tolyl)borate

A365515-1G A365515-25G A365515-5G
76724-676EA 19 USD
76724-676 76724-678 76724-680
Potassium(2-methylphenyl)trifluoroborate 96%
Potassium(2-methylphenyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₇H₇BF₃K
MW: 198.04
MDL Number: MFCD03790006
CAS Number: 274257-34-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR