Methyl-3-formyl-4-hydroxybenzoate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 3-formyl-4-hydroxybenzoate
A364546-100G
A364546-10G
A364546-1G
A364546-250MG
A364546-25G
A364546-5G
76724-648EA
1420.55
USD
76724-648
76724-650
76724-652
76724-654
76724-656
76724-658
Methyl-3-formyl-4-hydroxybenzoate 97%
Methyl-3-formyl-4-hydroxybenzoate
Precaución: for Research Use Only
Formula:
C₉H₈O₄ MW: 180.16 g/mol |
MDL Number:
MFCD01661473 CAS Number: 24589-99-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...