Methyl-4-(acetylamino)-2-hydroxybenzenecarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-acetamido-2-hydroxybenzoate

A364316-100G A364316-10G A364316-25G A364316-500G
76724-626EA 48.81 USD
76724-626 76724-628 76724-630 76724-632
Methyl-4-(acetylamino)-2-hydroxybenzenecarboxylate 97%
Methyl-4-(acetylamino)-2-hydroxybenzenecarboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₄
MW: 209.2 g/mol
MDL Number: MFCD00458326
CAS Number: 4093-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR