Methyl-3-cyclopentenecarboxylate 97%

Supplier: Ambeed
Danger

Synonyms: Methyl 3-cyclopentenecarboxylate

A363588-1G A363588-5G A363588-10G
76724-616EA 6.37 USD
76724-616 77594-076 77601-608
Methyl-3-cyclopentenecarboxylate 97%
Methyl-3-cyclopentenecarboxylate

Precaución: for Research Use Only

Formula: C₇H₁₀O₂
MW: 126.15 g/mol
MDL Number: MFCD04038661
CAS Number: 58101-60-3
UN: 2924
ADR: 3,8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR