N,N,N',N'-Tetrakis(4-methoxyphenyl)benzidine 98%

Supplier: Ambeed
Warning

A362464-1G A362464-250MG A362464-5G
76724-610EA 42 USD
76724-610 76724-612 76724-614
N,N,N',N'-Tetrakis(4-methoxyphenyl)benzidine 98%
N,N,N',N'-Tetrakis(4-methoxyphenyl)benzidine

Precaución: for Research Use Only

Formula: C₄₀H₃₆N₂O₄
MDL Number: MFCD09833415
CAS Number: 122738-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR