13-Bromo-1-tridecanol 95%

Supplier: Ambeed
Warning

Synonyms: 13-Bromotridecan-1-ol

A361373-100MG A361373-250MG A361373-25G A361373-5G A361373-1G
76724-602EA 24.41 USD
76724-602 76724-604 76724-606 76724-608 77601-602
13-Bromo-1-tridecanol 95%
13-Bromo-1-tridecanol

Precaución: for Research Use Only

Formula: HO(CH₂)₁₂CH₂Br
MW: 279.26 g/mol
MDL Number: MFCD00482035
CAS Number: 116754-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR