N-Boc-1,6-hexanediamine hydrochloride 98%

Supplier: Ambeed
Warning

A360427-100G A360427-25G A360427-5G
76724-582EA 442 USD
76724-582 76724-584 76724-586
N-Boc-1,6-hexanediamine hydrochloride 98%
N-Boc-1,6-hexanediamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₂₄N₂O₂·HCl
MDL Number: MFCD00039072
CAS Number: 65915-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR