4-Hydroxy-3-(1-phenylpropyl)-2H-chromen-2-one 98%

Supplier: Ambeed
Warning

A359847-100MG A359847-1G A359847-250MG A359847-5G
76724-576EA 66 USD
76724-576 76724-578 76724-580 77879-775
4-Hydroxy-3-(1-phenylpropyl)-2H-chromen-2-one 98%
4-Hydroxy-3-(1-phenylpropyl)-2H-chromen-2-one

Precaución: for Research Use Only

Formula: C₁₈H₁₆O₃
MDL Number: MFCD00865273
CAS Number: 435-97-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR