Propyl-4-hydroxybenzoate sodium salt 97%

Supplier: Ambeed
Warning

Synonyms: Propyl parahydroxybenzoate sodium salt, Sodium Propyl Parahydroxybenzoate, Propylparaben Sodium, Propylparaben sodium salt, 4-Hydroxybenzoic acid propyl ester sodium salt, Sodium 4-(propoxycarbonyl)phenolate, Sodium 4-propoxycarbonylphenoxide

A359800-100G A359800-10G A359800-25G A359800-500G
76724-572EA 20.16 USD
76724-572 76724-574 76840-428 77601-590
Propyl-4-hydroxybenzoate sodium salt 97%
Propyl-4-hydroxybenzoate sodium salt

Precaución: for Research Use Only

Formula: C₁₀H₁₁NaO₃
MW: 202.19 g/mol
MDL Number: MFCD00016477
CAS Number: 35285-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR