N,N-Diethylformamide 99%, Analytical reagent (AR)

Supplier: Ambeed
Danger

A357538-10G A357538-25G A357538-100G
76724-536EA 5 USD
76724-536 76724-538 77594-062
N,N-Diethylformamide 99%, Analytical reagent (AR)
N,N-Diethylformamide

Precaución: for Research Use Only

Formula: C₅H₁₁NO
MDL Number: MFCD00003287
CAS Number: 617-84-5
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR