Methyl-β-D-ribofuranoside 98% (α-isomer)
Supplier: Ambeed
|
Warning
|
Synonyms:
(2R,3S,4R,5R)-2-(Hydroxymethyl)-5-methoxytetrahydrofuran-3,4-diol, Methyl β-D-Ribofuranoside
A357535-1G
A357535-25G
A357535-5G
76724-530EA
9.55
USD
76724-530
76724-532
76724-534
Methyl-β-D-ribofuranoside 98% (α-isomer)
Methyl-beta-D-ribofuranoside
Precaución: for Research Use Only
Formula:
C₆H₁₂O₅ MW: 164.16 g/mol |
MDL Number:
MFCD00047075 CAS Number: 7473-45-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...