Methyl-2-furoate 95%

Supplier: Ambeed
Danger

Synonyms: Methyl 2-Furancarboxylate, Methyl furan-2-carboxylate, Methyl 2-furoate

A355718-25G A355718-100G
76724-522EA 5.31 USD
76724-522 77668-658
Methyl-2-furoate 95%
Methyl-2-furoate

Precaución: for Research Use Only

Formula: C₆H₆O₃
MW: 126.11 g/mol
MDL Number: MFCD00003236
CAS Number: 611-13-2
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR