N-Methyldipropylamine 98%

Supplier: Ambeed
Danger

Synonyms: N-Methyldi-n-propylamine

A354593-25G A354593-5G
76724-510EA 110.34 USD
76724-510 76724-512
N-Methyldipropylamine 98%
N-Methyldipropylamine

Precaución: for Research Use Only

Formula: C₇H₁₇N
MW: 115.22 g/mol
MDL Number: MFCD00048683
CAS Number: 3405-42-3
UN: 2924
ADR: 3,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR