3-[(Dimethylsilyl)oxy]-1,1,3,5,5-pentamethyltrisiloxane 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyltris(dimethylsiloxy)silane
A349303-100G
A349303-1G
A349303-25G
A349303-5G
76724-434EA
267.35
USD
76724-434
76724-436
76724-438
76724-440
3-[(Dimethylsilyl)oxy]-1,1,3,5,5-pentamethyltrisiloxane 96%
3-[(Dimethylsilyl)oxy]-1,1,3,5,5-pentamethyltrisiloxane
Precaución: for Research Use Only
Formula:
C₇H₂₄O₃Si₄ MW: 268.61 g/mol |
MDL Number:
MFCD00054789 CAS Number: 17082-46-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...