2-(2-Aminophenyl)benzimidazole 95%

Supplier: Ambeed
Warning

Synonyms: 2-(1H-Benzimidazol-2-yl)aniline, 2-(2-Aminophenyl)-1H-benzimidazole

A348459-100G A348459-1G A348459-25G A348459-5G
76724-420EA 455.13 USD
76724-420 76724-422 76724-424 76724-426
2-(2-Aminophenyl)benzimidazole 95%
2-(2-Aminophenyl)benzimidazole

Precaución: for Research Use Only

Formula: C₁₃H₁₁N₃
MW: 209.252 g/mol
MDL Number: MFCD00051496
CAS Number: 5805-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR