Methyl-2-fluoro-6-methoxybenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-fluoro-6-methoxybenzoate, 2-Fluoro-6-methoxybenzoic acid methyl ester

A346553-1G A346553-25G A346553-5G
76724-406EA 22.28 USD
76724-406 76724-408 76724-410
Methyl-2-fluoro-6-methoxybenzoate 98%
Methyl-2-fluoro-6-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉FO₃
MW: 184.17 g/mol
MDL Number: MFCD03788537
CAS Number: 178747-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR