Methyl 3-bromo-5-chlorobenzoate 98%

Supplier: Ambeed
Warning

A346014-100MG A346014-1G A346014-250MG
76724-400EA 6.37 USD
76724-400 76724-402 76724-404
Methyl 3-bromo-5-chlorobenzoate 98%
Methyl 3-bromo-5-chlorobenzoate

Precaución: for Research Use Only

Formula: C₈H₆BrClO₂
MW: 249.489 g/mol
MDL Number: MFCD07780734
CAS Number: 933585-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR