Methyl 4-ethylbenzoate 98%

Supplier: Ambeed
Warning

Synonyms: 4-Ethylbenzoic acid methyl ester

A344005-250MG A344005-25G A344005-5G A344005-1G
76724-376EA 12.74 USD
76724-376 76724-378 76724-380 77594-014
Methyl 4-ethylbenzoate 98%
Methyl 4-ethylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MW: 164.201 g/mol
MDL Number: MFCD00221477
CAS Number: 7364-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR