1-(Dimethylamino)-3-[2-(3-methoxyphenethyl)phenoxy]-2-propanol hydrochloride 97%

Supplier: Ambeed
Warning

A342548-100G A342548-10G A342548-1G A342548-25G A342548-5G
76724-366EA 427.99 USD
76724-366 76724-368 76724-370 76724-372 76724-374
1-(Dimethylamino)-3-[2-(3-methoxyphenethyl)phenoxy]-2-propanol hydrochloride 97%
1-(Dimethylamino)-3-[2-(3-methoxyphenethyl)phenoxy]-2-propanol hydrochloride

Precaución: for Research Use Only

Formula: C₂₀H₂₇NO₃
MDL Number: MFCD01697013
CAS Number: 135261-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR