Metyrapone 95%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-1,2-di(pyridin-3-yl)propan-1-one

A341725-100MG A341725-1G A341725-1MG A341725-250MG A341725-25MG A341725-5MG
76724-350EA 32 USD
76724-350 76724-352 76724-354 76724-356 76724-358 76724-360
Metyrapone 95%
Metyrapone

Precaución: for Research Use Only

Formula: C₁₄H₁₄N₂O
MDL Number: MFCD00006397
CAS Number: 54-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR