Ethyl-3-amino-1-methyl-1H-pyrazole-4-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-amino-1-methyl-1H-pyrazole-4-carboxylate

A339770-1G A339770-250MG A339770-5G
76724-320EA 47.75 USD
76724-320 76724-322 76724-324
Ethyl-3-amino-1-methyl-1H-pyrazole-4-carboxylate 95%
Ethyl-3-amino-1-methyl-1H-pyrazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₁N₃O₂
MW: 169.18 g/mol
MDL Number: MFCD00101268
CAS Number: 21230-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR