N-(2,6-Dichloro-4-methoxyphenyl)acetamide 95%

Supplier: Ambeed
Warning

A339489-100MG A339489-1G A339489-250MG
76724-306EA 105.03 USD
76724-306 76724-308 76724-310
N-(2,6-Dichloro-4-methoxyphenyl)acetamide 95%
N-(2,6-Dichloro-4-methoxyphenyl)acetamide

Precaución: for Research Use Only

Formula: C₉H₉Cl₂NO₂
MW: 234.08 g/mol
MDL Number: MFCD02090177
CAS Number: 136099-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR