Octafluoronaphthalene 92%

Supplier: Ambeed
Warning

Synonyms: Perfluoronaphthalene

A337387-100G A337387-1G A337387-250MG A337387-25G A337387-5G
76724-282EA 678.98 USD
76724-282 76724-284 76724-286 76724-288 76724-290
Octafluoronaphthalene 92%
Octafluoronaphthalene

Precaución: for Research Use Only

Formula: C₁₀F₈
MW: 272.1 g/mol
MDL Number: MFCD00014307
CAS Number: 313-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR